missing translation for 'onlineSavingsMsg'
Learn More

Heptanenitrile, 98%, Thermo Scientific Chemicals

Product Code. 11419443
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419443 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11419443 Supplier Thermo Scientific Alfa Aesar Supplier No. A12326.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-08-3 | Molecular Formula: C7H13N | Molecular Weight: 111.19 g/mol

The Heptanenitrile is used for producing 2, 3, 5, 6-tetrahexyl-pyrazine. As intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The Heptanenitrile is used for producing 2, 3, 5, 6-tetrahexyl-pyrazine. As intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Strong reducing agents, Strong acids, Strong bases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-08-3
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.19
MDL Number MFCD00039503
InChI Key SDAXRHHPNYTELL-UHFFFAOYSA-N
Synonym heptanonitrile, hexyl cyanide, enanthonitrile, 1-cyanohexane, 1-heptanonitrile, heptane-1-nitrile, n-heptanenitrile, unii-ua4z3qt98a, ua4z3qt98a, heptane nitrile
PubChem CID 12372
IUPAC Name heptanenitrile
SMILES CCCCCCC#N

Specifications

Melting Point -64°C
Density 0.81
Boiling Point 186°C to 187°C
Flash Point 58°C (136°F)
Refractive Index 1.415
Quantity 25 g
UN Number UN1992
Beilstein 1740405
Solubility Information Slightly soluble in water.
Formula Weight 111.19
Percent Purity 98%
Chemical Name or Material Heptanenitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.