missing translation for 'onlineSavingsMsg'
Learn More

Heptanophenone, 98+%, Thermo Scientific™

Product Code. 10551891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
10551891 25 g Glass bottle 25g
10202702 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10551891

Brand: Thermo Scientific Acros 279210250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1671-75-6
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.29
MDL Number MFCD00009539
InChI Key UXMQORVHJMUQFD-UHFFFAOYSA-N
Synonym heptanophenone, 1-heptanone, 1-phenyl, n-heptanophenone, unii-kx07wp06jy, kx07wp06jy, enanthophenone, hexyl phenyl ketone, 1-phenyl-heptan-1-one, 1-phenyl-1-heptanone #
PubChem CID 74282
IUPAC Name 1-phenylheptan-1-one
SMILES CCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 17°C
Color Yellow
Density 0.9400g/mL
Boiling Point 155°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5CO(CH2)5CH3
Refractive Index 1.5050 to 1.508
Quantity 25 g
Specific Gravity 0.94
Formula Weight 190.29
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Heptanophenone, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.