missing translation for 'onlineSavingsMsg'
Learn More

Heptanoyl chloride, 99%

Product Code. 11372096
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372096 250 g 250g
11362096 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372096 Supplier Thermo Scientific Alfa Aesar Supplier No. L03315.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2528-61-2 | C7H13ClO | 148.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2528-61-2
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD00000763
InChI Key UCVODTZQZHMTPN-UHFFFAOYSA-N
Synonym enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
PubChem CID 17313
IUPAC Name heptanoyl chloride
SMILES CCCCCCC(Cl)=O

Specifications

Melting Point -84°C
Density 0.96
Boiling Point 172°C to 174°C
Flash Point 58°C (136°F)
Odor Acrid
Linear Formula CH3(CH2)5COCl
Refractive Index 1.431
Quantity 250 g
UN Number UN2920
Beilstein 605636
Sensitivity Moisture sensitive
Formula Weight 148.63
Percent Purity 99%
Chemical Name or Material Heptanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.