missing translation for 'onlineSavingsMsg'
Learn More

Heptanoyl chloride, 99%

Product Code. 10761051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10761051 500 g Glass bottle 500g
10669014 25 g Glass bottle 25g
10502191 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10761051 Supplier Thermo Scientific Acros Supplier No. 164265000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2528-61-2 | C7H13ClO | 148.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2528-61-2
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD00000763
InChI Key UCVODTZQZHMTPN-UHFFFAOYSA-N
Synonym enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
PubChem CID 17313
IUPAC Name heptanoyl chloride
SMILES CCCCCCC(Cl)=O

Specifications

Melting Point -84°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 173°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5COCl
Refractive Index 1.4295 to 1.4315
Quantity 500 g
Beilstein 02, 340
Specific Gravity 0.96
Solubility Information Solubility in water: hydrolysis. Other solubilities: slightly soluble in carbontetrachloride,soluble in ethylether,ligroin
Formula Weight 148.63
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Heptanoyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.