missing translation for 'onlineSavingsMsg'
Learn More

Heptylbenzene, TRC

Product Code. 30403928
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403928 10 g -
30406005 25 g -
30395343 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403928 Supplier Toronto Research Chemicals Supplier No. TRCH28291010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1078-71-3
Molecular Formula C13 H20
Molecular Weight (g/mol) 176.3
Synonym Benzene, heptyl- (9CI, ACI), Heptane, 1-phenyl- (6CI, 7CI, 8CI), Heptylbenzene (ACI), 1-Heptylbenzene, 1-Phenylheptane, Heptane, 1(or 7)-phenyl-, n-Heptylbenzene, NSC 97413
IUPAC Name heptylbenzene
SMILES CCCCCCCc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H20/c1-2-3-4-5-7-10-13-11-8-6-9-12-13/h6,8-9,11-12H,2-5,7,10H2,1H3
Formula Weight 176.1565
Chemical Name or Material n-Heptylbenzene (Phenylheptane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.