missing translation for 'onlineSavingsMsg'
Learn More

Heptylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™

Product Code. 10535680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10535680 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10535680 Supplier Thermo Scientific Acros Supplier No. 433031000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13125-66-1 | C7H15BrMg | 203.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13125-66-1, 60-29-7
Color Brown
Density 0.8410g/mL
Boiling Point 35.0°C
Flash Point −30°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C7H15BrMg
Linear Formula CH3(CH2)6MgBr
MDL Number MFCD00013517
Quantity 100 mL
Specific Gravity 0.841
Synonym n-heptylmagnesium bromide, 1-heptylmagnesium bromide, heptyl magnesium bromide, magnesium, bromoheptyl, n-heptyl magnesium bromide, heptylmagnesium bromide solution, ldkgbijdngsvih-uhfffaoysa-m, 1-heptylmagnesium bromide 0.25 m in tetrahydrofuran, heptylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key LDKGBIJDNGSVIH-UHFFFAOYSA-M
SMILES CCCCCCC[Mg]Br
IUPAC Name magnesium;heptane;bromide
Molecular Weight (g/mol) 203.41
PubChem CID 10910616
Formula Weight 203.4
Physical Form Liquid
Chemical Name or Material Heptylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Heptylmagnesium bromide, 1M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.