Description
Upgrade your incubators to take advantage of O2 control.
Specifications
Specifications
| Description | 1 to 21% O2 control |
| For Use With (Equipment) | Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators |
| Lighting | LED |
| Oxygen Control | 1 to 21% |
| Type | O2 Control |