missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm CO2 Chamber 750 L

Product Code. 18784584 Shop All Thermo Scientific Products
Change view
Click to view available options
Door Style:
Glass door
Solid (outer), Glass (inner)
Solid outer door with inner glass door
Capacity (Metric):
247 L
750 L
Unit Size:
1 unit
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Capacity (Metric) Unit Size
18784584 Solid (outer), Glass (inner) 750 L 1 unit
18734594 Glass door 750 L 1 unit
31926565 Solid outer door with inner glass door 247 L Each
31926564 Glass door 247 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 18784584 Supplier Thermo Scientific™ Supplier No. ECS750
  • ONLINE EXCLUSIVE PRICE
    25426.50 €
    24563.00 € / Pack of 1
    Save 863.50 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Precisely control the essential parameters of your distinct cell growth or CO2 testing conditions — active humidity controls the incubation environment with accuracy.

Accurate control of three parameters - Temperature above and below ambient, relative humidity and CO2 - provide great environment in Cell growth and testing applications for reproducible results.

 

  • Uniform Temperature / CO2 / relative humidity throughout the chamber
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • CO2 control system ensures stable CO2 environment, range of 0 – 20 Vol-%.
  • Environmentally friendly water-efficient humidification system - with water tank or optional fixed water connection for in-house purified water
  • Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option.

Choose from a variety of country-specific sockets. (Ordered with separate order code)

TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 26.5 cu.ft.
Capacity (English) Shelf 66 lbs. (stationary)
Capacity (Metric) Shelf 30 kg (stationary)
Certifications/Compliance UL, CE
Data Outputs USB, Thermo Cloud via OPC UA
Capacity (Metric) 750 L
Chamber Material Stainless Steel (304/1.4301)
Material Powder-coated Galvanized Steel
No. of Shelves 3
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Type Growth Chamber
Display 7 in. Touchscreen user interface
Door Style Solid (outer), Glass (inner)
For Use With (Application) Cell growth, testing
Humidity Range 10 - 90% RH
Humidity Resolution 0.1% RH
Humidity Source Removable water tank
No. of Casters 4
Ports 2 access ports
Programmability Temperature, relative humidity, CO2 ramping
Shelf Area (English) 4.9 ft.2
Shelf Area (Metric) 0.46 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor - programmable
Warranty 24 months
Water Consumption <0.8 L per day - Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Water Resistivity 0.05 - 1 MΩ
Dimensions (L x W x H) 895 x 1080 x 1835 mm / 35.24 x 42.52 x 72.24 in.
Dimensions (LxWxH) Interior 630 x 840 x 1450 mm / 24.8 x 33.07 x 57.09 in.
Width (English) Shelf 31.3 in.
Width (Metric) Shelf 795 mm
Shipping Weight (English) 859 lbs.
Shipping Weight (Metric) 390 kg
Electrical Requirements 100/240 V, 50/60 Hz, 1400 W
Voltage 100/115/220/230 V
Wattage 1400 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.