missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Accurate control of three parameters - Temperature above and below ambient, relative humidity and CO2 - provide great environment in Cell growth and testing applications for reproducible results.
- Uniform Temperature / CO2 / relative humidity throughout the chamber
- Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant. CO2 control system ensures stable CO2 environment, range of 0 – 20 Vol-%.
- Environmentally friendly water-efficient humidification system - with water tank or optional fixed water connection for in-house purified water
- Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
- Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
- Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard.
- Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
- Simplified setup with no technician required; install using standard power cord.
- IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
- Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
- Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
- Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option.
Spécification
Spécification
| Alarm Types | Audible and Visible |
| Capacity (English) | 8.8 cu.ft. |
| Capacity (English) Shelf | 66.1 lb. |
| Capacity (Metric) Shelf | 30 kg |
| Certifications/Compliance | UL, CE |
| Data Outputs | USB, Thermo Cloud via OPC UA |
| Capacity (Metric) | 247 L |
| Chamber Material | Powder-coated Cold Rolled Steel |
| Lighting | No |
| Material | Powder-coated Cold Rolled Steel |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?