missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm Light Chambers 750 L

Product Code. 18714594 Shop All Thermo Scientific Products
Change view
Click to view available options
Door Style:
Glass door
Solid outer door with inner glass door
For Use With (Application):
ICH light testing, plant growth, light testing, animal hatching
Plant growth, light testing, animal hatching
Includes:
Light modules not included - ICH testing: 333979G01 / Standard: 333980G01
Light modules not included - for plant growth/testing/animal hatching: 333980G01
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style For Use With (Application) Includes Unit Size
18714594 Glass door Plant growth, light testing, animal hatching Light modules not included - for plant growth/testing/animal hatching: 333980G01 1 unit
18764584 Solid outer door with inner glass door ICH light testing, plant growth, light testing, animal hatching Light modules not included - ICH testing: 333979G01 / Standard: 333980G01 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 18714594 Supplier Thermo Scientific™ Supplier No. ELG750
  • ONLINE EXCLUSIVE PRICE
    24728.03 €
    23370.00 € / Pack of 1
    Save 1358.03 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Accurate and even light distribution with Long-lasting LED lighting for (ICH) light testing, plant growth and animal hatching — relative humidity and temperature are controlled to meet specified environment.

Accurate control of three parameters - Temperature above and below ambient, relative humidity and light - provide great environment in testing applications and for biological research with plants and animals. Choice of ICH module provides capability for efficient ICH light testing in Pharma.

  • Uniform Temperature / relative humidity / light exposure for samples.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • LED light system provides specified light exposure on 1, 2 or 3 shelves; programmable to adapt light quality / intensity and on/off timing - e.g. for day-night cycles. Choose from two light modules (ordered separately):
    • 1. ICH module, includes UV LED for ICH light testing;can only be used in ELS750 - model with solid outer door - for safety reasons
    • 2. Testing module for plant growth / daylight testing / animal hatching
  • Environmentally friendly water-efficient humidification system - with water tank or optional fix water connection for in-house purified water
  • Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard (with Connect Edge solution)
  • Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Exterior glass door also available (not for ICH module use - for safety reasons).
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 26.5 cu.ft
Capacity (English) Shelf 66 lbs (stationary)
Capacity (Metric) Shelf 30 kg (stationary)
Certifications/Compliance UL, CE
Data Outputs USB, Thermo Cloud via OPC UA
Capacity (Metric) 750 L
Chamber Material Stainless Steel (304/1.4301)
Material Powder-coated Galvanized Steel
No. of Shelves 3
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Temperature Range (Metric) 15 to 45°C with light; 5 to 60°C without light
Type Vertical Light Chamber
Display 7 in. Touchscreen user interface
Door Style Glass door
For Use With (Application) Plant growth, light testing, animal hatching
Humidity Range 10 - 80% RH with light; 10 -90% RH w/o light
Humidity Resolution 0.1% RH
Humidity Source Removable water tank
Includes Light modules not included - for plant growth/testing/animal hatching: 333980G01
No. of Casters 4
Ports 2 access ports
Programmability Temperature, relative humidity, light intensity
Shelf Area (English) 4.9 ft2

Shelf Area (Metric) 0.46 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor - programmable
Temperature Resolution 0.1°C
Temperature Stability ≤ ±0.3°C @ 25°C to 40°C
Temperature Uniformity ≤ ±2.0°C @ 25°C to 40°C
Warranty 24 months
Water Consumption 0.2- 1.5 L per day; Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Water Resistivity 0.05 - 1 MΩ
Dimensions (L x W x H) 895 x 1080 x 1835 mm / 35.24 x 42.52 x 72.24 in
Dimensions (LxWxH) Interior 630 x 840 x 1450 mm / 24.8 x 33.07 x 57.09 in
Length (Metric) Shelf 795 mm
Width (English) Shelf 31.3 in.
Shipping Weight (English) 842 lbs
Shipping Weight (Metric) 382 kg
Electrical Requirements 100 - 240V, 50-60Hz, 1700W
Voltage 100/240 V
Wattage 1700 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.