missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm™ IMP400 Refrigerated Incubator, 381 L, Stainless Steel

Product Code. 15618540 Shop All Thermo Scientific Products
Change view
Click to view available options
Voltage:
100/240 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
15618540 100/240 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15618540 Supplier Thermo Scientific™ Supplier No. 51031565
  • ONLINE EXCLUSIVE PRICE
    12276.08 €
    11600.00 € / Pack of 1
    Save 676.08 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Temperature Range: 5°C to 70°C

Achieve untapped potential in incubation with the temperature precision you need for applications that demand it.. With a temperature range of 5-70°C, Thermo Scientific Heratherm Refrigerated Incubators are equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results. With no hazardous refrigerants, these units use less energy than the conventional product, making it the ideal choice for energy-conscious labs

  • Temperature range +5 to +70°C
  • Available in 2 sizes: benchtopmodel 178L / 6.3 cuft and a floormodel with casters 381L / 13.5 cu.ft.
  • Great temperature uniformity and stability to keep the samples in a safe environment –> uniformity as low as +/-0.3°C (20°C), stability as low as +/-0.1°C
  • Energy consumption as low as 80Wh/h (bench top) / 150Wh/h(floor) at 20°C - Note that comparable compressor cooled units consume up to 6 times this amount*.
  • CFC free / HFC free polyurethane insulation foam for great temperature performance and condensate prevention
  • Heated door with intelligent energy control to ensure no condensation – even at low temperatures
  • Sophisticated timer function with weekly, daily or real time mode
  • Intuitive programmability function for defined temperature ramping, soak/cool and dwell: store up to 10 programs with 10 easy steps
  • Adjustable over temperature shut-off for protocols that require setting of over temperature for protection – (class 2 according to norm DIN12880)
  • Safe viewing of samples through internal glass door with limited impact on temperature
  • Safe containment with automatic over and under temperature alarm
  • Standard access port for data monitoring (large diameter of 42mm / 1.65 in.)
  • Flexible shelf system for optimal loading, easy to remove for cleaning, with non-tip protection
  • Stainless steel interior (1.4301 / ASTM 304) and rounded corners for easy cleaning
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 13.5 cu. ft.
Certifications/Compliance CE, CSA
Description Floor Model
Capacity (Metric) 381 L
Chamber Material Stainless Steel (304/1.4301)
Dimensions (D x W) Shelf 20.79 x 21.06 in. (528 x 498 mm)
Material Coated Steel
No. of Shelves 2 std./17 max.
Plug Type N. America: Nema 5-15 plug w/ 120V 60Hz config.; All others: CEE7/7 with 230V 50Hz config.
Temperature Range (Metric) 5°C to 70°C
Type Automated Incubator
Display Large Vacuum Fluorescent
Door Style Solid outer door & internal glass door
Insulation CFC free/HFC free polyurethane insulation foam
Model IMP400
No. of Doors 1
Port Size 1.6 in. (42mm)
Refrigerant Peltier technology: no refrigerant
Temperature Accuracy 0.1°C
Temperature Uniformity ≤0.3°C at 20°C; ≤0.7°C at 37°C
Dimensions (D x W x H) Interior 524 x 544 x 1335 mm (20.6 x 21.4 x 52.6 in.)
Dimensions (L x W x H) 770 x 778 x 1545 mm (30.3 x 230.6 x 60.8 in.)
Height (English) 60.8 in. (casters not included: 7.1 in.)
Height (Metric) 1545 mm (casters not included: 180 mm)
Shipping Weight (English) 476 lb.
Shipping Weight (Metric) 216 kg
Weight (English) 368 lb.
Weight (Metric) 167 kg
Electrical Requirements 100/240 V, 50/60 Hz, 3.8-9.0 A, 900 w
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.