missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm Stability Chambers

Product Code. 31926572 Shop All Thermo Scientific Products
Change view
Click to view available options
Door Style:
Glass
Glass door
Solid (outer), Glass (inner)
Solid outer door with inner glass door
Capacity (Metric):
247 L
750 L
Unit Size:
1 unit
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Capacity (Metric) Unit Size
31926572 Glass door 247 L Each
18724594 Solid (outer), Glass (inner) 750 L 1 unit
18704594 Glass 750 L 1 unit
31926570 Solid outer door with inner glass door 247 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31926572 Supplier Thermo Scientific™ Supplier No. ESG250
  • ONLINE EXCLUSIVE PRICE
    17550.00 €
    15803.19 € / Pack of 1
    Save 1746.81 €
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gain confidence in your product when testing stability and shelf life with robust Heratherm Stability chambers designed to meet a variety of quality control demands.

Temperature and humidity controls, well suited for stability testing, shelf life testing, and environmental testing.

  • Broad relative humidity range to cover many applications, including humidity levels below ambient.
  • Environmentally friendly water-efficient humidification system.
  • Minimal condensate in standard applications.
  • Integrated water tank; fixed water connection for in-house purified water option available.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 8.8 cu.ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Thermo Cloud via OPC UA
Capacity (Metric) 247 L
Chamber Material Powder-coated Cold Rolled Steel
Lighting No
Material Powder-coated Cold Rolled Steel
No. of Shelves 2
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Temperature Range (Metric) 0 to 70°C
Type Stability Chamber
Display 7 in. Touchscreen user interface
Door Style Glass door
For Use With (Application) Stability testing, environmental testing, storage
Humidity Range 10 to 90% RH
Humidity Source Removable water tank
No. of Casters None
Ports 2 access ports
Programmability Temperature and humidity ramping
Shelf Area (English) 3.05 ft.2
Shelf Area (Metric) 0.28 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor - programmable
Temperature Resolution 0.1°C
Temperature Stability ≤ ±0.1°C @ 20°C to 37°C
Temperature Uniformity ≤ ±0.5°C @ 20°C to 37°C
Warranty 24 months
Water Consumption ≤ 0.25 L per day - Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Water Resistivity 0.05 - 1 MΩ
Width (English) 35.5 in.
Width (English) Interior 26 in.
Width (English) Shelf 24.2 in.
Width (Metric) 901 mm
Width (Metric) Interior 660 mm
Width (Metric) Shelf 615 mm
Height (English) 31.5 in
Height (Metric) 1128 mm
Height (Metric) Interior 800 mm
Depth (English) 31.1 in.
Depth (English) Interior 20.4 in.
Depth (Metric) 790 mm
Depth (Metric) Interior 519 mm
Shipping Weight (English) 448 lb.
Shipping Weight (Metric) 203 kg
Electrical Requirements 100 /240 V, 50/60 Hz
Voltage 100/115/220/230 V
Wattage 900 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.