missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hesperidin

Product Code. 16451509
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451509 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451509 Supplier MedChemExpress Supplier No. HY15337
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hesperidin (Hesperetin 7-rutinoside), a flavanone glycoside, is isolated from citrus fruits. Hesperidin has numerous biological properties, such as decreasing inflammatory mediators and exerting significant antioxidant effects. Hesperidin also exhibits antitumor and antiallergic activities.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 610.56 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 520-26-3
Product Type Natural Products
Purity 99.19%
Solubility DMSO : ≥ 33 mg/mL (54.05 mM)
Synonym Hesperetin 7-rutinoside
Target Apoptosis∣Autophagy∣Reactive Oxygen Species∣Endogenous Metabolite
Quantity 1 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.