missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals HEV IgM ELISA 3.0 (CE) 96 wells

Product Code. 30564756 Shop All MP Biomedicals Products
Change view
Click to view available options
Target Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species
30564756 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30564756 Supplier MP Biomedicals Supplier No. 23160096
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

HEV IgM ELISA 3.0 (CE) is an enzyme linked immunosorbent assay intended for the detection of IgM antibodies to Hepatitis E Virus (HEV) in human serum or plasma.

The wells of the polystyrene microplate strips are coated with highly conserved conformational ORF2 epitope. Human serum or plasma, diluted in diluent buffer, are incubated in these coated wells. HEV specific antibodies, if present, will bind to the antigens immobilized on the solid phase. After incubation, the wells are thoroughly washed to remove unbound materials and mouse monoclonal anti-human IgM antibodies labeled with horseradish peroxidase is added to the wells. This labeled antibody will bind to any antigen-antibody complexes previously formed and excess unbound labeled antibodies are removed by washing. A colourless substrate solution containing 3,3',5,5'-tetramethylbenzidine (TMB) is then added to each well. The presence of specific antibodies is indicated by the presence of a blue colour after incubation, which changes to yellow when the colour reaction is terminated by the addition of acid. The intensity of the resulting yellow product is measured at 450 nm using a spectrophotometer and is proportional to the amount of antibodies present in the specimen.

  • CE-marked
  • 96 wells
  • For professional use only
  • For HEV antibodies detection

Specifications

Detectable Analytes Hepatitis E Virus (HEV)
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Type ELISA Kit
Sample Type Human Serum, Human Plasma
Target Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.