Learn More
Hexa-n-butylditin, 97%
Description
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Spezifikation
Spezifikation
| Density | 1.152 |
| Boiling Point | 197°C to 198°C (10 mmHg) |
| Flash Point | 124°C (255°F) |
| Linear Formula | (CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3 |
| Refractive Index | 1.512 |
| Quantity | 10 g |
| UN Number | UN2788 |
| Beilstein | 3605476 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Immiscible with water. |
| Mehr anzeigen |
Sicherheit und Handhabung
- Hexa-n-butylditin
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Hexabutyldistannane
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.