missing translation for 'onlineSavingsMsg'
Learn More

Hexa-n-butylditin, 97%

Product Code. 10072031 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10072031 10 g Glass bottle 10g
10246011 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072031 Supplier Thermo Scientific Acros Supplier No. 351510100
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 813-19-4 | C24H54Sn2 | 580.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-19-4
Molecular Formula C24H54Sn2
Molecular Weight (g/mol) 580.12
MDL Number MFCD00009417
InChI Key REDSKZBUUUQMSK-UHFFFAOYSA-N
Synonym hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
PubChem CID 6327815
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC

Specifications

Color Colorless to Yellow
Density 1.1480g/mL
Boiling Point 197.0°C to 198.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ([CH3(CH2)3]3Sn-)2
Refractive Index 1.5100 to 1.5130
Quantity 10 g
Specific Gravity 1.148
Solubility Information Solubility in water: insoluble
Formula Weight 580.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Hexa-n-butylditin
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexa-n-butylditin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.