missing translation for 'onlineSavingsMsg'
Learn More

Hexaamminecobalt(III) chloride, 99.999%, (trace metal basis)

Product Code. 10481811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10481811 5 g Glass bottle 5g
10134070 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10481811 Supplier Thermo Scientific Acros Supplier No. 192870050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10534-89-1 | H18Cl3CoN6 | 267.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 10534-89-1
Molecular Formula H18Cl3CoN6
Molecular Weight (g/mol) 267.47
MDL Number MFCD00036304

Specifica

Melting Point 217°C
Color Orange to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999%
Linear Formula [Co(NH3)6]Cl3
Merck Index 15, 4710
Solubility Information Solubility in water: soluble
Formula Weight 267.47
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Hexaamminecobalt(III) chloride, 99.999%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.