missing translation for 'onlineSavingsMsg'
Learn More

Hexaammineruthenium(III) chloride, Ru 32.1% min

Product Code. 11346018
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346018 0.25 g 0.25g
11366018 1 g 1g
11356018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346018 Supplier Thermo Scientific Alfa Aesar Supplier No. 010511.01
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14282-91-8 | Cl3H18N6Ru | 309.61 g/mol

Depending on the choice of suitable reductive or oxidative quencher this catalyst can be used to trigger photoreduction or photooxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14282-91-8
Molecular Formula Cl3H18N6Ru
Molecular Weight (g/mol) 309.61
MDL Number MFCD00011478
InChI Key GBDZMMXUOBAJMN-UHFFFAOYSA-K
Synonym hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride
PubChem CID 159731
SMILES N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Color Yellow
Quantity 0.25 g
UN Number UN3288
Solubility Information Very soluble in water.
IUPAC Name ruthenium(3+) hexaamine trichloride
Formula Weight 309.61
Concentration or Composition (by Analyte or Components) Ru 32.1% min.
Physical Form Powder
Chemical Name or Material Hexaammineruthenium(III) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.