missing translation for 'onlineSavingsMsg'
Learn More

Hexaammineruthenium(III) chloride, Ru 32.1% min

Product Code. 11366018
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11366018

Brand: Thermo Scientific Alfa Aesar 010511.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14282-91-8 | Cl3H18N6Ru | 309.61 g/mol

Depending on the choice of suitable reductive or oxidative quencher this catalyst can be used to trigger photoreduction or photooxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14282-91-8
Molecular Formula Cl3H18N6Ru
Molecular Weight (g/mol) 309.61
MDL Number MFCD00011478
InChI Key GBDZMMXUOBAJMN-UHFFFAOYSA-K
Synonym hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride
PubChem CID 159731
SMILES N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Color Yellow
Quantity 1 g
UN Number UN3288
Solubility Information Very soluble in water.
IUPAC Name azane;ruthenium(3+);trichloride
Formula Weight 309.61
Concentration or Composition (by Analyte or Components) Ru 32.1% min.
Physical Form Powder
Chemical Name or Material Hexaammineruthenium(III) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.