missing translation for 'onlineSavingsMsg'
Learn More

Hexacarbonylchromium, 99%

Product Code. 11392148
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
Product Code. Quantity unitSize
11392148 10 g 10g
11382148 1 g 1g
11372148 50 g 50g
3 options
This item is not returnable. View return policy

Product Code. 11392148

Brand: Thermo Scientific Alfa Aesar 013061.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13007-92-6 | C6CrO6 | 220.056 g/mol

Hexacarbonylchromium is widely used as a catalyst for isomerization of alkenes. It is involved in the hydrogenation of 1,3-conjugated dienes and oxidation of allylic and benzylic positions. It acts as a precursor to prepare phenanthrenes, tricarbonyl(arene)chromium and Fischer carbine complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13007-92-6
Molecular Formula C6CrO6
Molecular Weight (g/mol) 220.056
MDL Number MFCD00010945
InChI Key KOTQLLUQLXWWDK-UHFFFAOYSA-N
Synonym chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g
PubChem CID 25589
ChEBI CHEBI:33031
IUPAC Name carbon monoxide;chromium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Specifications

Melting Point ∽152°C (decomposition)
Color White
Physical Form Crystals
Quantity 10 g
Assay Percent Range 99%
Solubility Information Soluble in ether,chloroform,tetrahydropyran and methylene chloride. Insoluble in water.
Formula Weight 220.06
Sensitivity Light sensitive
Vapor Pressure 0.35 hPa
Density 1.77
Chemical Name or Material Hexacarbonylchromium
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexacarbonylchromium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H350i-May cause cancer by inhalation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Hexacarbonylchromium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.