Learn More
Hexacarbonyltungsten, 97%
Description
Hexacarbonyltungsten is used as a precursor to a Fischer carbine complex, which used in the hiyama cross-coupling reactions. It is widely used in electron beam-induced deposition technique as well as a source of tungsten atoms. It is utilized to desulfurize organosulfur compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 172°C to 174°C |
| Color | White |
| Density | 2.65 |
| Assay Percent Range | 97% |
| Odor | Nearly odorless |
| Quantity | 10 g |
| Solubility Information | Slightly soluble in tetrahydrofuran. Insoluble in water. |
| Formula Weight | 351.91 |
| Vapor Pressure | 1.596hPa (1mm Hg) at 67°C |
| Physical Form | Crystalline |
| Mehr anzeigen |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.