missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroethane, 99%

Product Code. 10143720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143720 100 g Plastic bottle 100g
10522161 1 kg Plastic bottle 1kg
10487160 2.5 kg Plastic bottle 2.5kg
11994471 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10143720 Supplier Thermo Scientific Acros Supplier No. 148541000
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 67-72-1 | C2Cl6 | 236.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-72-1
Molecular Formula C2Cl6
Molecular Weight (g/mol) 236.72
MDL Number MFCD00000799
InChI Key VHHHONWQHHHLTI-UHFFFAOYSA-N
Synonym hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin
PubChem CID 6214
ChEBI CHEBI:39227
SMILES ClC(Cl)(Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 184.0°C to 190.0°C
Color White
Density 2.0910g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula Cl3CCCl3
Quantity 100 g
Beilstein 01,87
Merck Index 15,4715
Specific Gravity 2.091
Solubility Information Solubility in water: 0.05g/L (22°C). Other solubilities: soluble in most common organic solvents,>50g/100mL ethanol
IUPAC Name hexachloroethane
Formula Weight 236.74
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Hexachloroethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexachloroethane
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.