missing translation for 'onlineSavingsMsg'
Learn More

Hexacosanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10740751
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
506-46-7
C26H52O2
396.7
MFCD00002811
XMHIUKTWLZUKEX-UHFFFAOYSA-N
cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
10469
CHEBI:31009
hexacosanoic acid
CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
This item is not returnable. View return policy
506-46-7
C26H52O2
396.7
MFCD00002811
XMHIUKTWLZUKEX-UHFFFAOYSA-N
cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
10469
CHEBI:31009
hexacosanoic acid
CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 87.0°C to 89.0°C
Color White
Density 0.8100g/mL
Boiling Point 250.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)24COOH
Refractive Index 1.4301
Quantity 100 mg
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether and acetone
Formula Weight 396.7
Percent Purity 98%
Physical Form Shiny Fluffy Crystalline Powder
Chemical Name or Material Hexacosanoic acid, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'