missing translation for 'onlineSavingsMsg'
Learn More

Hexacosanoic acid, 98%

Product Code. p-141614 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 10740751

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 506-46-7 | C26H52O2 | 396.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-46-7
Molecular Formula C26H52O2
Molecular Weight (g/mol) 396.7
MDL Number MFCD00002811
InChI Key XMHIUKTWLZUKEX-UHFFFAOYSA-N
Synonym cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
PubChem CID 10469
ChEBI CHEBI:31009
IUPAC Name hexacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 87.0°C to 89.0°C
Color White
Density 0.8100g/mL
Boiling Point 250.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)24COOH
Refractive Index 1.4301
Quantity 100 mg
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether and acetone
Formula Weight 396.7
Percent Purity 98%
Physical Form Shiny Fluffy Crystalline Powder
Chemical Name or Material Hexacosanoic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.