missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane, Extra Pure, SLR, Fisher Chemical

Product Code. 10204580 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Amber glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10204580 100 mL Amber glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10204580 Supplier Fisher Chemical Supplier No. H/0270/05
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Specifications

CAS 544-76-3
Melting Point 18°C
Color Colorless
Boiling Point 287°C
Packaging Amber glass bottle
Molecular Formula C16H34
MDL Number 8998
Quantity 100 mL
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC
IUPAC Name hexadecane
Molecular Weight (g/mol) 226.448
PubChem CID 11006
ChEBI CHEBI:45296
Formula Weight 226.44g/mol
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Hexadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.