missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-15,15,16,16,16-d5 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-15,15,16,16,16-D5 Acid |
| CAS | 285979-77-3 |
| Molecular Formula | C16 D5 H27 O2 |
| Quantity | 0.25 g |
| Synonym | Hexadecanoic-15,15,16,16,16-d5 acid (9 CI), Hexadecanoic-15,15,16,16,16-D5 Acid, Palmitic acid-15,15,16,16,16-d5, [15,15,16,16,16-2H5]Hexadecanoic acid, [15,15,16,16,16-2H5]Palmitic acid, Hexadecanoic Acid-15,15,16,16,16-d5 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCCCCCC(=O)O |
| IUPAC Name | 15,15,16,16,16-pentadeuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 261.4549 |
| Formula Weight | 261.2716 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction