missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic Acid-1,2-13C2, CDN
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-1,2-13C2 Acid |
| CAS | 86683-25-2 |
| Molecular Formula | 13C2 C14 H32 O2 |
| Quantity | 0.5 g |
| Synonym | Hexadecanoic-1,2-13C2 acid (9 CI), Palmitic acid-1,2-13C2, [1,2-13C2]Hexadecanoic acid, [1,2-13C2]Palmitic acid, Hexadecanoic Acid-1,2-13C2 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i15+1,16+1 |
| SMILES | CCCCCCCCCCCCCC[13 CH2][13 C](=O)O |
| IUPAC Name | (1,2-13C2)hexadecanoic acid |
| Molecular Weight (g/mol) | 258.4094 |
| Formula Weight | 258.2469 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?