missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyl Methacrylate, TRC

Product Code. 30407706
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407706 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407706 Supplier Toronto Research Chemicals Supplier No. TRCH2938001G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexadecyl Methacrylate
CAS 2495-27-4
Molecular Formula C20H38O2
Quantity 1 g
Synonym 2-Methyl-2-propenoic Acid Hexadecyl Ester, Methacrylic Acid Hexadecyl Ester, 1-Hexadecanol Methacrylate, Blemmer CMA, Cetyl Methacrylate, Hexadecyl 2-Methyl-2-propenoate, Palmityl Methacrylate
InChI Formula InChI=1S/C20H38O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-20(21)19(2)3/h2,4-18H2,1,3H3
SMILES CCCCCCCCCCCCCCCCOC(=O)C(=C)C
IUPAC Name hexadecyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 310.51
Formula Weight 310.2872
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.