missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Bromide, 99+%

Product Code. 10645852 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic Drum
Unit Size:
100g
10g
10kg
2.5kg
25g
500g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10645852 100 g Plastic Bottle 100g
10368600 10 g Glass Bottle 10g
10246520 25 g Plastic Bottle 25g
10541671 500 g Plastic Bottle 500g
11621789 2.5 kg Plastic Drum 2.5kg
13448488 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10645852 Supplier Thermo Scientific Acros Supplier No. 227161000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 57-09-0 | C19H42BrN | 364.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
PubChem CID 5974
ChEBI CHEBI:3567
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Physical Form Crystalline Powder
Melting Point 230.0°C
Decomposition Information >230°C
Quantity 100 g
Flash Point 244°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 99+%
Linear Formula CH3(CH2)15N(CH3)3Br
Beilstein 04, IV, 819
Fieser 15,77
Merck Index 15, 2024
Synonym Cetrimonium Bromide, Cetyltrimethylammonium Bromide, CTABr
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone
IUPAC Name hexadecyltrimethylazanium bromide
Formula Weight 364.45
Percent Purity ≥99%
Chemical Name or Material Hexadecyltrimethylammonium Bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexadecyltrimethylammonium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.