missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Bromide, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15518794 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15518794 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518794 Supplier Avantor J.T.Baker Supplier No. N12107

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 57-09-0
Flash Point 244.00°C
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C19H42BrN
MDL Number MFCD00011772
Quantity 500 g
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name hexadecyl(trimethyl)azanium;bromide
Molecular Weight (g/mol) 364.46
PubChem CID 5974
ChEBI CHEBI:3567
Formula Weight 364.46
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Hexadecyltrimethylammonium Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.