missing translation for 'onlineSavingsMsg'
Learn More

Hexaethylphosphorus Triamide, 95%

Product Code. 10686984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686984 100 g Glass bottle 100g
10662075 5 g Glass bottle 5g
10085840 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10686984 Supplier Thermo Scientific Supplier No. 423081000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2283-11-6 | C12H30N3P | 247.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2283-11-6
Molecular Formula C12H30N3P
Molecular Weight (g/mol) 247.37
MDL Number MFCD00009041
InChI Key FDIOSTIIZGWENY-UHFFFAOYSA-N
Synonym tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine
PubChem CID 75292
IUPAC Name [bis(diethylamino)phosphanyl]diethylamine
SMILES CCN(CC)P(N(CC)CC)N(CC)CC

Specifications

Color Colorless to Orange-Yellow
Density 0.8900g/mL
Boiling Point 111°C (6.0 mmHg)
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ((C2H5)2N)3P
Refractive Index 1.4720 to 1.475
Quantity 100 g
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble
Formula Weight 247.36
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Hexaethylphosphorus triamide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexaethylphosphorus triamide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.