missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroacetone Hydrate, TRC

Product Code. 30405948
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405948 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405948 Supplier Toronto Research Chemicals Supplier No. TRCH3051302.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Hexafluoroacetone Hydrate
CAS 10057-27-9
Molecular Formula C3 F6 O . xH2O
Quantity 2.5 g
InChI Formula InChI=1S/C3F6O.H2O/c4-2(5,6)1(10)3(7,8)9;/h;1H2
SMILES O=C(C(F)(F)F)C(F)(F)F.O
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one hydrate
Molecular Weight (g/mol) 184.03
Formula Weight 183.9959
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.