missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroglutaric acid, 97%, ACROS Organics™

Product Code. 10073660 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10073660

Brand: Thermo Scientific Acros 306600050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 376-73-8
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C5H2F6O4
MDL Number MFCD00004172
Quantity 5 g
Synonym hexafluoroglutaric acid, perfluoroglutaric acid, hexafluoropentanedioic acid, pentanedioic acid, hexafluoro, glutaric acid, hexafluoro, pentanedioic acid, 2,2,3,3,4,4-hexafluoro, acmc-209iut, perfluoropentanedioic acid, glutaric acid, hexafluoro-8ci
InChI Key CCUWGJDGLACFQT-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O
IUPAC Name hexafluoropentanedioic acid
Molecular Weight (g/mol) 240.06
PubChem CID 67827
Formula Weight 240.06
Chemical Name or Material Hexafluoroglutaric acid, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.