missing translation for 'onlineSavingsMsg'
Learn More

Hexahydro-1,3,5-triphenyl-1,3,5-triazine, 98%, Thermo Scientific™

Product Code. 11382167
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382167 10 g 10g
11392167 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11382167 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L09620.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 91-78-1
Molecular Formula C21H21N3
Molecular Weight (g/mol) 315.42
MDL Number MFCD00043590
InChI Key VASMRQAVWVVDPA-UHFFFAOYSA-N
Synonym hexahydro-1,3,5-triphenyl-1,3,5-triazine, 1,3,5-triazine, hexahydro-1,3,5-triphenyl, anhydroformaldehyde aniline, 1,3,5-triphenylhexahydro-1,3,5-triazine, 1,3,5-triphenylhexahydro-s-triazine, unii-x3tz97444k, s-triazine, hexahydro-1,3,5-triphenyl, 1,3,5-triphenyl-1,3,5-triazaperhydroine, anhydroformaldehyde aniline van, methyleneaniline trimer
PubChem CID 7065
IUPAC Name 1,3,5-triphenyl-1,3,5-triazinane
SMILES C1N(CN(CN1C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4

Tekniske data

Melting Point 190°C to 192°C
Quantity 10 g
Beilstein 91510
Formula Weight 315.42
Percent Purity 98%
Chemical Name or Material Hexahydro-1,3,5-triphenyl-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.