missing translation for 'onlineSavingsMsg'
Learn More

Hexahydro-1,3-isobenzofurandione, TRC

Product Code. 30138068
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
Unit Size:
100g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138068 25 g 25g
30132448 100 g 100g
17922325 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138068 Supplier Toronto Research Chemicals Supplier No. TRCH29402025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-42-7
Molecular Formula C8 H10 O3
Molecular Weight (g/mol) 154.16
Synonym 1,3-Isobenzofurandione, hexahydro-, 1,2-Cyclohexanedicarboxylic anhydride (6CI,8CI), Hexahydro-1,3-isobenzofurandione, 1,2-Cyclohexanedicarboxylic acid anhydride, Aradur HY 925, Araldite HT 904, Araldite HT 907, Araldite HY 907, Araldite HY 925, Araldite Hardener HY 925, Cyclohexane-1,2-dicarboxylic acid anhydride, Cyclohexanedicarboxylic anhydride, EP 400B, Epilox H 11-01, HHPA, HT 907, HV 136, HY 925, Hardener HY 925, Hexahydro-2-benzofuran-1,3-dione, Hexahydrophthalic acid anhydride, Hexahydrophthalic anhydride, Lekutherm Hardener H, MH 700E, NSC 8622, Rikacid HH, Rikacid HH-A, Rikacid HHPA, Rikacid MH 600, Rikacid MH 700E, Rutadur AG, Rutapox HX
IUPAC Name 3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2CCCCC12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H10O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h5-6H,1-4H2
Formula Weight 154.063
Chemical Name or Material Hexahydro-1,3-isobenzofurandione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.