missing translation for 'onlineSavingsMsg'
Learn More

Hexahydro-4-methylphthalic Anhydride, TRC

Product Code. 30397021
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397021 5 g 5g
30402388 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397021 Supplier Toronto Research Chemicals Supplier No. TRCH2937855G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19438-60-9
Molecular Formula C9 H12 O3
Molecular Weight (g/mol) 168.19
Synonym 1,3-Isobenzofurandione, hexahydro-5-methyl-, 1,2-Cyclohexanedicarboxylic anhydride, 4-methyl- (8CI), Hexahydro-5-methyl-1,3-isobenzofurandione, 1-Methylcyclohexane-4,5-Dicarboxylic anhydride, 4-Methyl-1,2-cyclohexanedicarboxylic anhydride, 4-Methylhexahydrophthalic acid anhydride, 4-Methylhexahydrophthalic anhydride, HN 7000, Hexahydro-4-methylphthalic anhydride, Lindride 52, NSC 128883, Quinhard HH 400, Rikacid MH, Rikacid MH-T, WH 700
IUPAC Name 5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1CCC2C(C1)C(=O)OC2=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H12O3/c1-5-2-3-6-7(4-5)9(11)12-8(6)10/h5-7H,2-4H2,1H3
Formula Weight 168.0786
Chemical Name or Material 4-Methyl-1,2-cyclohexanedicarboxylic acid anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.