missing translation for 'onlineSavingsMsg'
Learn More

gamma-Hexalactone, 98%

Product Code. 15580662 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15580662 100 g 100g
15595086 25 g 25g
15505096 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15580662 Supplier Thermo Scientific Supplier No. 394171000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 695-06-7 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-06-7
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005401
InChI Key JBFHTYHTHYHCDJ-UHFFFAOYNA-N
Synonym gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one
PubChem CID 12756
ChEBI CHEBI:85235
IUPAC Name 5-ethyloxolan-2-one
SMILES CCC1CCC(=O)O1

Specifications

Melting Point -18.0°C
Density 1.0230g/mL
Boiling Point 220.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 100 g
Specific Gravity 1.023
Solubility Information Solubility in water: slightly soluble.
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material γ-Hexalactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.