missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylcyclotrisiloxane, 98%

Product Code. 15129243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15129243 500 g Glass Bottle 500g
10617182 25 g Glass Bottle 25g
10750721 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15129243 Supplier Thermo Scientific Acros Supplier No. 216465000
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 541-05-9 | C6H18O3Si3 | 222.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-05-9
Molecular Formula C6H18O3Si3
Molecular Weight (g/mol) 222.46
MDL Number MFCD00005943
InChI Key HTDJPCNNEPUOOQ-UHFFFAOYSA-N
Synonym hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185
PubChem CID 10914
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Melting Point 63.0°C to 67.0°C
Color Colorless to White
Boiling Point 134.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 500 g
UN Number 1325
Beilstein 04,III,1884
Merck Index 09,4559
IUPAC Name hexamethyl-1,3,5,2,4,6-trioxatrisilinane
Formula Weight 222.47
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material Hexamethylcyclotrisiloxane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Hexamethylcyclotrisiloxane
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.