missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilane, 98+%

Product Code. 11474534
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474534 25 g 25g
11464534 5 g 5g
11484534 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474534 Supplier Thermo Scientific Alfa Aesar Supplier No. A13155.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1450-14-2 | C6H18Si2 | 146.38 g/mol

Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.

Solubility
Insoluble in water. Soluble in alcohol, ether and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-14-2
Molecular Formula C6H18Si2
Molecular Weight (g/mol) 146.38
MDL Number MFCD00008258
InChI Key NEXSMEBSBIABKL-UHFFFAOYSA-N
Synonym hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane
PubChem CID 74057
IUPAC Name trimethyl(trimethylsilyl)silane
SMILES C[Si](C)(C)[Si](C)(C)C

Specifications

Boiling Point 112°C to 113°C
Melting Point 12°C to 13°C
Quantity 25 g
UN Number UN1993
Beilstein 1633463
Flash Point −1°C (30°F)
Solubility Information Insoluble in water. Soluble in alcohol,ether and acetone.
Formula Weight 146.38
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.422
Density 0.721
Chemical Name or Material Hexamethyldisilane
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.