missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylditin, 99%

Product Code. 10723914 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10723914

Brand: Thermo Scientific Acros 295850250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 661-69-8 | C6H18Sn2 | 327.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-69-8
Molecular Formula C6H18Sn2
Molecular Weight (g/mol) 327.63
MDL Number MFCD00008277
InChI Key CCRMAATUKBYMPA-UHFFFAOYSA-N
Synonym hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci
PubChem CID 6327618
SMILES C[Sn](C)(C)[Sn](C)(C)C

Specifications

Melting Point 23.0°C to 24.0°C
Color Brown
Boiling Point 182.0°C (756.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SnSn(CH3)3
Refractive Index 1.5370 to 1.5410
Quantity 25 g
Beilstein 04, IV, 4346
Fieser 17,143
IUPAC Name trimethyltin
Formula Weight 327.63
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Hexamethylditin, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexamethylditin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.