missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyleneimine, 98+%

Product Code. 11416916
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416916 500 mL 500mL
11406916 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416916 Supplier Thermo Scientific Alfa Aesar Supplier No. B24007.AP
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-49-9 | C6H13N | 99.177 g/mol

Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-49-9
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00006934
InChI Key ZSIQJIWKELUFRJ-UHFFFAOYSA-N
Synonym hexamethyleneimine, homopiperidine, hexahydroazepine, hexahydro-1h-azepine, perhydroazepine, 1h-azepine, hexahydro, hexamethylenimine, azacycloheptane, azepan, hexamethylene imine
PubChem CID 8119
ChEBI CHEBI:32616
IUPAC Name azepane
SMILES C1CCCNCC1

Specifications

Melting Point -37°C
Density 0.88
Boiling Point 138°C to 139°C
Flash Point 22°C (71°F)
Odor Ammonia-like
Refractive Index 1.466
Quantity 500 mL
UN Number UN2493
Beilstein 1084
Solubility Information Fully miscible in water.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material Hexamethyleneimine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.