missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, 99+%

Product Code. 11443437
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443437 5000 g 5000g
11433437 250 g 250g
11423437 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443437 Supplier Thermo Scientific Alfa Aesar Supplier No. A17213.0I
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-97-0 | C6H12N4 | 140.19 g/mol

Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

Solubility
Soluble in water, ethanol, chloroform and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Avoid conditions like flames, sparks and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
MDL Number MFCD00006895
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Melting Point ∼280°C (sublimation)
Density 1.33
Flash Point 250°C (482°F)
Quantity 5000 g
UN Number UN1328
Beilstein 2018
Sensitivity Hygroscopic
Merck Index 14,5966
Solubility Information Soluble in water,ethanol,chloroform and acetone.
Formula Weight 140.19
Percent Purity ≥99%
Chemical Name or Material Hexamethylenetetramine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexamethylenetetramine
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.