missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, ACS, 99+%

Product Code. 11377918
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377918 500 g 500g
11387918 100 g 100g
11367918 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377918 Supplier Thermo Scientific Alfa Aesar Supplier No. 036462.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 100-97-0 | C6H12N4 | 140.19 g/mol

Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
MDL Number MFCD00006895
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Melting Point 280°C (sublimation)
Color White
Density 1.33 g/mL
Flash Point 250°C (482°F)
Quantity 500 g
UN Number UN1328
Beilstein 2018
Sensitivity Hygroscopic
Merck Index 14,5966
Solubility Information Soluble in water,dimethyl sulfoxide,chloroform,ethanol and acetone. Slightly soluble in ether.
Formula Weight 140.19
Percent Purity ≥99%
Grade ACS
Physical Form Solid
Chemical Name or Material Hexamethylenetetramine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexamethylenetetramine
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/face, hands and anyexposed skin.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: 1,3,5,7-Tetraazatricyclo[3.3.1.1(3,7)]decane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.