missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, TRC

Product Code. 17958839
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17958839 50 mg 50mg
17967929 10 g 10g
17903945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17958839 Supplier Toronto Research Chemicals Supplier No. TRCH29422050MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6 H12 N4
Molecular Weight (g/mol) 140.19
Synonym Methenamine, 1,3,5,7-Tetraazotricyclo[3.3.1.13,7]decane, Hexamethylenetetramine (8CI), 1,3,5,7-Tetraazaadamantane, 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane (Urotropine), Aceto HMT, Aminoform, Ammoform, Antihydral, Cohedur H 30, Cystogen, Duirexol, EL 10, EL 10 (corrosion inhibitor), Ekagom H, Formamine, Formin, Formin (heterocycle), HA, HMT, HMTA, Heksa K, Herax UTS, Heterin, Hexa, Hexa (vulcanization accelerator), Hexa B, Hexa-Flo-Pulver, Hexaform, Hexamethylenamine, Hexamine, Hexamine (heterocycle), Hexamine Superfine, Hexasan, Methenamide, Methenamin, Methenamine, NSC 26346, NSC 403347, Nocceler H, Nocceler H-PO, Preparation AF, Rhenogran HEXA 80, S 4, S 4 (heterocycle), Sanceler H, Sanceler HT, Sanceler HT-PO, Thixon 715B, Uramin, Uratrine, Uritone, Urodeine, Urotropin, Urotropine, Vulkacit H, Vulkacit H 30, Xametrin
IUPAC Name 1,3,5,7-Tetraazotricyclo[3.3.1.13,7]decane
SMILES C1N2CN3CN(CN1C3)C2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2
Formula Weight 140.1062
Chemical Name or Material Urotropine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.