missing translation for 'onlineSavingsMsg'
Learn More

Hexan-1-ol, Extra Pure, SLR, Fisher Chemical

Product Code. 10766481 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10766481 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10766481 Supplier Fisher Chemical Supplier No. H/0450/PB17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 111-27-3
Melting Point -52°C
Color Colorless
Boiling Point 156°C
Packaging Plastic Coated Glass bottle
Molecular Formula C6H14O
MDL Number 2982
Quantity 2.5 L
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILES CCCCCCO
IUPAC Name hexan-1-ol
Molecular Weight (g/mol) 102.177
PubChem CID 8103
ChEBI CHEBI:87393
Formula Weight 102.18g/mol
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.