missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Hexane

Product Code. 16633954
Change view
Click to view available options
Quantity:
7 L
18 L
Unit Size:
18 L
7 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16633954 18 L 18 L
15945460 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 16633954 Supplier Riedel-de Haën Supplier No. 3229318L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Hexane, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-Hexane
IUPAC Name hexane
SMILES CCCCCC

Specifications

pH Not applicable
Melting Point -95°C
Color Colorless
Density 0.660 to 0.668 g/cm3 (20°C)
Quantity 18 L
Flash Point 7.4 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula CH3(CH2)4CH3
Vapor Pressure 160 hPa (20°C)
Viscosity 0.31 mPa.s (20°C)
UN Number UN1208
Beilstein 1730733
Boiling Point 66°C to 69°C (1.013 hPa)
Percent Purity ≥99.00%
Grade ACS
Chemical Name or Material Hexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.