missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, CHROMASOLV™, ≥97%, LC-MS, Honeywell Riedel-de Haën™
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
Specifications
| Density | 0.659g/mL at 25°C |
| Quantity | 4 x 2.5 L |
| Explosion Limit | 0.077 |
| Flash Point | −26°C (−14.8°F) |
| Ignition Point | 453°F |
| Linear Formula | CH3(CH2)4CH3 |
| Vapor Pressure | 5.2 psi (37.7°C), 132 mmHg (20°C), 256 mmHg (37.7°C) |
| UN Number | UN1208 |
| Formula Weight | 86.18 |
| Percent Purity | ≥97% |
| Show More |
Safety and Handling
Product Identifier
- Hexane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : n-hexane
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?