missing translation for 'onlineSavingsMsg'
Learn More

Hexane, For pesticide residue analysis, Honeywell Riedel-de Haën™

Product Code. p-7072099
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Unit Size:
1 L
2.5 L
Pack of 4
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15694190

Brand: Honeywell-Chromasolv 344841L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.659g/mL at 25°C
Quantity 1 L
Explosion Limit 0.077
Flash Point −26°C (−14.8°F)
Ignition Point 453°F
Linear Formula CH3(CH2)4CH3
Vapor Pressure 5.2 psi (37.7°C), 256 mmHg (37.7°C), 132 mmHg (20°C)
UN Number UN1208
Beilstein 1730733
Formula Weight 86.18g/mol
Grade CHROMASOLV
Chemical Name or Material Hexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.