missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Mixture Of Isomers, ≥98.5%, ACS Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™

Product Code. 15611500
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
Unit Size:
2 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15611500

Brand: Honeywell Riedel-de Haën 1561752L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

ACS Spectrophotometric Grade,≥98.5%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -154°C
CAS 110-54-3
Appearance Liquid
Density 0.672g/mL at 25°C
Boiling Point 48°C to 65°C
Quantity 2 L
Explosion Limit 0.075
Flash Point −23°C (−9.4°F)
Ignition Point 437°F
Packaging Glass Bottle
Refractive Index 1.379
Molecular Formula C6H14
MDL Number MFCD02179311
UN Number UN1208
Beilstein 1730733
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
PubChem CID 8058
ChEBI CHEBI:29021
Formula Weight 86.18g/mol
Percent Purity ≥98.5%
Grade ACS
Chemical Name or Material Hexane, Mixture Of Isomers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.