missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanedioic-d8 Acid Dihydrazide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Adipic Acid Dihydrazide-D8 |
| CAS | 1202865-37-9 |
| Molecular Formula | C6 D8 H6 N4 O2 |
| Quantity | 0.1 g |
| Synonym | 2,2,3,3,4,4,5,5-Octadeuteriohexanedihydrazide, Hexanedioic-d8 Acid Dihydrazide, Hexanedioic Acid Dihydrazide-D8, Adipic-d8 acid dihydrazide, Adipic Acid Dihydrazide-D8 |
| InChI Formula | InChI=1 S/C6H14N4O2/c7-9-5(11)3-1-2-4-6(12)10-8/h1-4,7-8H2,(H,9,11)(H,10,12)/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(C(=O)NN)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)NN |
| IUPAC Name | 2,2,3,3,4,4,5,5-octadeuteriohexanedihydrazide |
| Molecular Weight (g/mol) | 182.2503 |
| Formula Weight | 182.1619 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction